1,2,3-Trimethoxy-5-(prop-2-yn-1-yloxy)benzene, tech

Lieferant: Apollo Scientific

OR32363-1G
APOSOR32363-1GEA 1570 EUR
APOSOR32363-1G
1,2,3-Trimethoxy-5-(prop-2-yn-1-yloxy)benzene, tech
1,2,3-Trimethoxy-5-(prop-2-yn-1-yloxy)benzene
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05975060
CAS-Nummer: 860787-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR