Methyl 1-methyl-4-(3-methylbutanoyl)-1H-pyrrole-2-carboxylate, tech

Lieferant: Apollo Scientific

OR32365-1G
APOSOR32365-1GEA 1570 EUR
APOSOR32365-1G
Methyl 1-methyl-4-(3-methylbutanoyl)-1H-pyrrole-2-carboxylate, tech
Methyl 1-methyl-4-(3-methylbutanoyl)-1H-pyrrole-2-carboxylate
Formel: C₁₂H₁₇NO₃
Molecular Weight: 223,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16089977
CAS-Nummer: 1211020-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR