6-Ethylpyridine-2-carbonitrile 97%

Lieferant: Apollo Scientific

OR32366-1G
APOSOR32366-1GEA 1570 EUR
APOSOR32366-1G
6-Ethylpyridine-2-carbonitrile 97%
6-Ethylpyridine-2-carbonitrile
Formel: C₈H₈N₂
Molecular Weight: 132,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21604038
CAS-Nummer: 59146-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR