1-[2-(1H-Imidazol-1-yl)phenyl]ethan-1-one, tech

Lieferant: Apollo Scientific

OR32368-1G
APOSOR32368-1GEA 1570 EUR
APOSOR32368-1G
1-[2-(1H-Imidazol-1-yl)phenyl]ethan-1-one, tech
1-[2-(1H-Imidazol-1-yl)phenyl]ethan-1-one
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127221
CAS-Nummer: 20513-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR