N4,N4-Dimethyl-5-phenylpyrimidine-2,4-diamine 97%

Lieferant: Apollo Scientific

OR32369-1G
APOSOR32369-1GEA 874 EUR
APOSOR32369-1G
N4,N4-Dimethyl-5-phenylpyrimidine-2,4-diamine 97%
N4,N4-Dimethyl-5-phenylpyrimidine-2,4-diamine
Formel: C₁₂H₁₄N₄
Molecular Weight: 214.266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127225
CAS-Nummer: 320424-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR