(4Z)-4-[(Dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one 95%
Lieferant: Apollo Scientific
OR32370-1G
APOSOR32370-1GEA
1120
EUR
APOSOR32370-1G
(4Z)-4-[(Dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one 95%
(4Z)-4-[(Dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Formel:
C₁₃H₁₅N₃O Molecular Weight: 229,278 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100702 CAS-Nummer: 65644-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...