3-(2-Chlorophenyl)-5-methyl-1,2-oxazole-4-carbonitrile, tech

Lieferant: Apollo Scientific

OR32371-1G
APOSOR32371-1GEA 1570 EUR
APOSOR32371-1G
3-(2-Chlorophenyl)-5-methyl-1,2-oxazole-4-carbonitrile, tech
3-(2-Chlorophenyl)-5-methyl-1,2-oxazole-4-carbonitrile
Formel: C₁₁H₇ClN₂O
Molecular Weight: 218,639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214710
CAS-Nummer: 320424-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR