(1E,4E)-1-(Dimethylamino)-5-(4-methoxyphenyl)penta-1,4-dien-3-one 95%

Lieferant: Apollo Scientific

OR32376-1G
APOSOR32376-1GEA 427 EUR
APOSOR32376-1G
(1E,4E)-1-(Dimethylamino)-5-(4-methoxyphenyl)penta-1,4-dien-3-one 95%
(1E,4E)-1-(Dimethylamino)-5-(4-methoxyphenyl)penta-1,4-dien-3-one
Formel: C₁₄H₁₇NO₂
Molecular Weight: 231.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172121
CAS-Nummer: 373650-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR