1-[(1-Phenyl-1H-1,2,4-triazol-3-yl)oxy]propan-2-one, tech
Lieferant: Apollo Scientific
OR32377-1G
APOSOR32377-1GEA
1570
EUR
APOSOR32377-1G
1-[(1-Phenyl-1H-1,2,4-triazol-3-yl)oxy]propan-2-one, tech
1-[(1-Phenyl-1H-1,2,4-triazol-3-yl)oxy]propan-2-one
Formel:
C₁₁H₁₁N₃O₂ Molecular Weight: 217,224 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138721 CAS-Nummer: 320416-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...