(2E)-3-(Dimethylamino)-2-(4-methoxyphenyl)prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR32378-1G
APOSOR32378-1GEA 1570 EUR
APOSOR32378-1G
(2E)-3-(Dimethylamino)-2-(4-methoxyphenyl)prop-2-enenitrile, tech
(2E)-3-(Dimethylamino)-2-(4-methoxyphenyl)prop-2-enenitrile
Formel: C₁₂H₁₄N₂O
Molecular Weight: 202,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138583
CAS-Nummer: 36758-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR