(3E)-1-(3,4-Dichlorophenyl)-3-(methoxyimino)propan-1-one, tech

Lieferant: Apollo Scientific

OR32379-1G
APOSOR32379-1GEA 1570 EUR
APOSOR32379-1G
(3E)-1-(3,4-Dichlorophenyl)-3-(methoxyimino)propan-1-one, tech
(3E)-1-(3,4-Dichlorophenyl)-3-(methoxyimino)propan-1-one
Formel: C₁₀H₉Cl₂NO₂
Molecular Weight: 246,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127284
CAS-Nummer: 320416-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR