7-Phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32381-1G
APOSOR32381-1GEA 1570 EUR
APOSOR32381-1G
7-Phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
7-Phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formel: C₁₃H₈N₄
Molecular Weight: 220,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138880
CAS-Nummer: 72860-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR