7-(4-Bromophenyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32382-1G
APOSOR32382-1GEA 0 EUR
APOSOR32382-1G
7-(4-Bromophenyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
7-(4-Bromophenyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formel: C₁₃H₇BrN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00138886
CAS-Nummer: 320417-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR