7-[(Z)-2-Phenylethenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32383-1G
APOSOR32383-1GEA 1570 EUR
APOSOR32383-1G
7-[(Z)-2-Phenylethenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
7-[(Z)-2-Phenylethenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formel: C₁₅H₁₀N₄
Molecular Weight: 246,267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138888
CAS-Nummer: 320417-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR