4'-(2-Chlorethyl)acetophenon 95%

Lieferant: Apollo Scientific

Synonyme: 1-[4-(2-Chloroethyl)phenyl]ethan-1-one

OR32384-25G OR32384-5G
APOSOR32384-25GEA 887 EUR
APOSOR32384-25G APOSOR32384-5G
4'-(2-Chlorethyl)acetophenon 95%
4'-(2-Chlorethyl)acetophenon
Formel: C₁₀H₁₁ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00016350
CAS-Nummer: 69614-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR