1-(4-Chlorophenyl)-2-(1H-imidazol-1-yl)ethan-1-one, tech

Lieferant: Apollo Scientific

OR32385-1G
APOSOR32385-1GEA 1210 EUR
APOSOR32385-1G
1-(4-Chlorophenyl)-2-(1H-imidazol-1-yl)ethan-1-one, tech
1-(4-Chlorophenyl)-2-(1H-imidazol-1-yl)ethan-1-one
Formel: C₁₁H₉ClN₂O
Molecular Weight: 220,655 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215352
CAS-Nummer: 24155-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR