1-(6-Chloropyridin-2-yl)-1H-pyrazol-5-ol, tech

Lieferant: Apollo Scientific

OR32386-1G
APOSOR32386-1GEA 1570 EUR
APOSOR32386-1G
1-(6-Chloropyridin-2-yl)-1H-pyrazol-5-ol, tech
1-(6-Chloropyridin-2-yl)-1H-pyrazol-5-ol
Formel: C₈H₆ClN₃O
Molecular Weight: 195,606 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02185866
CAS-Nummer: 1007030-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR