Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%

Lieferant: Apollo Scientific

OR32387-1G
APOSOR32387-1GEA 671 EUR
APOSOR32387-1G
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate
Formel: C₁₂H₉ClN₂O₃
Molecular Weight: 264,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141141
CAS-Nummer: 129109-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR