Methyl 1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32390-1G
APOSOR32390-1GEA 1570 EUR
APOSOR32390-1G
Methyl 1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech
Methyl 1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate
Formel: C₁₄H₁₂ClNO₃
Molecular Weight: 277.703 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140817
CAS-Nummer: 320419-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR