1-(3-Chlorophenyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one, tech

Lieferant: Apollo Scientific

OR32391-1G
APOSOR32391-1GEA 1570 EUR
APOSOR32391-1G
1-(3-Chlorophenyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one, tech
1-(3-Chlorophenyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one
Formel: C₁₂H₈ClN₃O
Molecular Weight: 245,664 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140663
CAS-Nummer: 320419-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR