(3Z)-3-(3,4-Dimethoxyphenyl)-4-(dimethylamino)but-3-en-2-one, tech

Lieferant: Apollo Scientific

OR32393-1G
APOSOR32393-1GEA 1570 EUR
APOSOR32393-1G
(3Z)-3-(3,4-Dimethoxyphenyl)-4-(dimethylamino)but-3-en-2-one, tech
(3Z)-3-(3,4-Dimethoxyphenyl)-4-(dimethylamino)but-3-en-2-one
Formel: C₁₄H₁₉NO₃
Molecular Weight: 249,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173441
CAS-Nummer: 76512-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR