O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 2-[(Ammoniooxy)methyl]-1,4-dichlorobenzene chloride

OR32398-1G
APOSOR32398-1GEA 671 EUR
APOSOR32398-1G
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride 95%
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride
Formel: C₇H₇Cl₂NO·HCl
Molecular Weight: 228,504 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01114576
CAS-Nummer: 317821-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR