3-(4-Methoxyphenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

OR32400-1G
APOSOR32400-1GEA 1570 EUR
APOSOR32400-1G
3-(4-Methoxyphenoxy)butan-2-one, tech
3-(4-Methoxyphenoxy)butan-2-one
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231719
CAS-Nummer: 24264-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR