1-(Dimethylamino)-4-(3-pyridinyloxy)-1-penten-3-one 95%

Lieferant: Apollo Scientific

OR32401-1G
APOSOR32401-1GEA 270 EUR
APOSOR32401-1G
1-(Dimethylamino)-4-(3-pyridinyloxy)-1-penten-3-one 95%
1-(Dimethylamino)-4-(3-pyridinyloxy)-1-penten-3-one
Formel: C₁₂H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08056611
CAS-Nummer: 886361-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR