1-Methyl-N'-phenyl-1H-imidazole-2-carbohydrazide, tech

Lieferant: Apollo Scientific

OR32402-1G
APOSOR32402-1GEA 1570 EUR
APOSOR32402-1G
1-Methyl-N'-phenyl-1H-imidazole-2-carbohydrazide, tech
1-Methyl-N'-phenyl-1H-imidazole-2-carbohydrazide
Formel: C₁₁H₁₂N₄O
Molecular Weight: 216,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794988
CAS-Nummer: 400078-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR