(2E)-1-[4-(Benzenesulphinyl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech
Lieferant: Apollo Scientific
OR32404-1G
APOSOR32404-1GEA
1570
EUR
APOSOR32404-1G
(2E)-1-[4-(Benzenesulphinyl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-[4-(Benzenesulphinyl)phenyl]-3-(dimethylamino)prop-2-en-1-one
Formel:
C₁₇H₁₇NO₂S Molecular Weight: 299,387 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664260 CAS-Nummer: 321432-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...