(3E)-2-(Morpholin-4-yl)-4-phenylbut-3-enenitrile, tech

Lieferant: Apollo Scientific

OR32405-1G
APOSOR32405-1GEA 1570 EUR
APOSOR32405-1G
(3E)-2-(Morpholin-4-yl)-4-phenylbut-3-enenitrile, tech
(3E)-2-(Morpholin-4-yl)-4-phenylbut-3-enenitrile
Formel: C₁₄H₁₆N₂O
Molecular Weight: 228,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173087
CAS-Nummer: 101132-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR