4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-5-methoxy-2-(pyridin-2-yl)pyrimidine, 4-Chloro-5-methoxy-2-(2-pyridyl)pyrimidine

OR32406-1G
APOSOR32406-1GEA 671 EUR
APOSOR32406-1G
4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine 95%
4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine
Formel: C₁₀H₈ClN₃O
Molecular Weight: 221,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215412
CAS-Nummer: 321432-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR