(2E)-1-(3-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32407-1G
APOSOR32407-1GEA 1570 EUR
APOSOR32407-1G
(2E)-1-(3-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-(3-Chlorothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one
Formel: C₉H₁₀ClNOS
Lagertemperatur: Raumtemperatur
MDL: MFCD00793820
CAS-Nummer: 166196-77-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR