2-(2-Chlorobenzoyl)-1H-imidazole, tech

Lieferant: Apollo Scientific

OR32408-1G
APOSOR32408-1GEA 1570 EUR
APOSOR32408-1G
2-(2-Chlorobenzoyl)-1H-imidazole, tech
2-(2-Chlorobenzoyl)-1H-imidazole
Formel: C₁₀H₇ClN₂O
Molecular Weight: 206.628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00125193
CAS-Nummer: 287196-66-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR