(1E)-1-(Dimethylamino)-4-(4-methylphenoxy)pent-1-en-3-one, tech

Lieferant: Apollo Scientific

OR32410-1G
APOSOR32410-1GEA 0 EUR
APOSOR32410-1G
(1E)-1-(Dimethylamino)-4-(4-methylphenoxy)pent-1-en-3-one, tech
(1E)-1-(Dimethylamino)-4-(4-methylphenoxy)pent-1-en-3-one
Formel: C₁₄H₁₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00243598
CAS-Nummer: 477864-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR