(1E)-4-(2-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech

Lieferant: Apollo Scientific

OR32411-1G
APOSOR32411-1GEA 1570 EUR
APOSOR32411-1G
(1E)-4-(2-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech
(1E)-4-(2-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one
Formel: C₁₃H₁₆ClNO₂
Molecular Weight: 253,725 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243621
CAS-Nummer: 1164521-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR