Methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32412-1G
APOSOR32412-1GEA 1570 EUR
APOSOR32412-1G
Methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate
Formel: C₇H₉NO₃
Molecular Weight: 155,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244305
CAS-Nummer: 77867-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR