Methyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Lieferant: Apollo Scientific
OR32413-1G
APOSOR32413-1GEA
1570
EUR
APOSOR32413-1G
Methyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate
Formel:
C₁₀H₁₄N₂O₃ Molecular Weight: 210,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00244306 CAS-Nummer: 477865-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...