(2E)-3-Amino-3-(4-phenylpiperazin-1-yl)prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR32415-1G
APOSOR32415-1GEA 671 EUR
APOSOR32415-1G
(2E)-3-Amino-3-(4-phenylpiperazin-1-yl)prop-2-enenitrile, tech
(2E)-3-Amino-3-(4-phenylpiperazin-1-yl)prop-2-enenitrile
Formel: C₁₃H₁₆N₄
Molecular Weight: 228,293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793935
CAS-Nummer: 141991-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR