(2E)-3-Amino-3-(pyrrolidin-1-yl)prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR32416-1G
APOSOR32416-1GEA 1570 EUR
APOSOR32416-1G
(2E)-3-Amino-3-(pyrrolidin-1-yl)prop-2-enenitrile, tech
(2E)-3-Amino-3-(pyrrolidin-1-yl)prop-2-enenitrile
Formel: C₇H₁₁N₃
Molecular Weight: 137,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793938
CAS-Nummer: 72139-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR