4-Chloro-2-(4-methoxyphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32417-1G
APOSOR32417-1GEA 1570 EUR
APOSOR32417-1G
4-Chloro-2-(4-methoxyphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech
4-Chloro-2-(4-methoxyphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one
Formel: C₁₄H₁₄ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00793839
CAS-Nummer: 477867-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR