(3Z)-3-[2-(4-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one, tech

Lieferant: Apollo Scientific

OR32418-1G
APOSOR32418-1GEA 1570 EUR
APOSOR32418-1G
(3Z)-3-[2-(4-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one, tech
(3Z)-3-[2-(4-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one
Formel: C₁₂H₁₅N₃O
Molecular Weight: 217,267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794636
CAS-Nummer: 3464-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR