3-Brom-5-chlor-2-methoxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR3242-1G
APOSOR3242-1GEA
21.9
EUR
APOSOR3242-1G
3-Brom-5-chlor-2-methoxypyridin
3-Bromo-5-chloro-2-methoxypyridine
Formel:
C₆H₅BrClNO Schmelzpunkt: 48…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059322 CAS-Nummer: 102830-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...