Ethyl 2-[(4-methylphenyl)formamido]acetate, tech

Lieferant: Apollo Scientific

OR32423-1G
APOSOR32423-1GEA 1570 EUR
APOSOR32423-1G
Ethyl 2-[(4-methylphenyl)formamido]acetate, tech
Ethyl 2-[(4-methylphenyl)formamido]acetate
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793748
CAS-Nummer: 122081-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR