Ethyl 2-[(3-methylphenyl)formamido]acetate, tech

Lieferant: Apollo Scientific

OR32424-1G
APOSOR32424-1GEA 1570 EUR
APOSOR32424-1G
Ethyl 2-[(3-methylphenyl)formamido]acetate, tech
Ethyl 2-[(3-methylphenyl)formamido]acetate
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221.252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793749
CAS-Nummer: 330466-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR