1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone 95%

Lieferant: Apollo Scientific

OR32427-1G
APOSOR32427-1GEA 270 EUR
APOSOR32427-1G
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone 95%
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone
Formel: C₁₁H₁₂BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00793408
CAS-Nummer: 338392-04-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR