6-Methyl-4-oxo-1-[4-(propan-2-yl)phenyl]-1,4-dihydropyridine-3-carboxylic acid, tech

Lieferant: Apollo Scientific

OR32428-1G
APOSOR32428-1GEA 1570 EUR
APOSOR32428-1G
6-Methyl-4-oxo-1-[4-(propan-2-yl)phenyl]-1,4-dihydropyridine-3-carboxylic acid, tech
6-Methyl-4-oxo-1-[4-(propan-2-yl)phenyl]-1,4-dihydropyridine-3-carboxylic acid
Formel: C₁₆H₁₇NO₃
Molecular Weight: 271,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794140
CAS-Nummer: 477868-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR