1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(pyrrolidin-1-yl)propan-2-ol, tech

Lieferant: Apollo Scientific

OR32430-1G
APOSOR32430-1GEA 1570 EUR
APOSOR32430-1G
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(pyrrolidin-1-yl)propan-2-ol, tech
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(pyrrolidin-1-yl)propan-2-ol
Formel: C₁₆H₂₂N₂O₂
Molecular Weight: 274,358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315608
CAS-Nummer: 338392-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR