1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(morpholin-4-yl)propan-2-ol, tech

Lieferant: Apollo Scientific

OR32431-1G
APOSOR32431-1GEA 1570 EUR
APOSOR32431-1G
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(morpholin-4-yl)propan-2-ol, tech
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(morpholin-4-yl)propan-2-ol
Formel: C₁₆H₂₂N₂O₃
Molecular Weight: 290,357 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315611
CAS-Nummer: 338392-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR