1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(piperidin-1-yl)propan-2-ol, tech
Lieferant: Apollo Scientific
OR32432-1G
APOSOR32432-1GEA
1570
EUR
APOSOR32432-1G
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(piperidin-1-yl)propan-2-ol, tech
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(piperidin-1-yl)propan-2-ol
Formel:
C₁₇H₂₄N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315615 CAS-Nummer: 338392-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...