N-(2,4,6-Trichloro-3-hydroxyphenyl)acetamide, tech

Lieferant: Apollo Scientific

OR32435-1G
APOSOR32435-1GEA 1570 EUR
APOSOR32435-1G
N-(2,4,6-Trichloro-3-hydroxyphenyl)acetamide, tech
N-(2,4,6-Trichloro-3-hydroxyphenyl)acetamide
Formel: C₈H₆Cl₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18429805
CAS-Nummer: 7397-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR