N-tert-Butylmorpholine-4-carbothioamide, tech

Lieferant: Apollo Scientific

OR32436-1G
APOSOR32436-1GEA 1570 EUR
APOSOR32436-1G
N-tert-Butylmorpholine-4-carbothioamide, tech
N-tert-Butylmorpholine-4-carbothioamide
Formel: C₉H₁₈N₂OS
Molecular Weight: 202,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24391363
CAS-Nummer: 14294-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR