4-[(1E)-(Methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol, tech

Lieferant: Apollo Scientific

OR32437-1G
APOSOR32437-1GEA 1570 EUR
APOSOR32437-1G
4-[(1E)-(Methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol, tech
4-[(1E)-(Methoxyimino)methyl]-1-methyl-3-phenyl-1H-pyrazol-5-ol
Formel: C₁₂H₁₃N₃O₂
Molecular Weight: 231.251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315295
CAS-Nummer: 321533-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR