Ethyl 2-[(3E)-2-oxo-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-3-ylidene]acetate, tech

Lieferant: Apollo Scientific

OR32438-1G
APOSOR32438-1GEA 1570 EUR
APOSOR32438-1G
Ethyl 2-[(3E)-2-oxo-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-3-ylidene]acetate, tech
Ethyl 2-[(3E)-2-oxo-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-3-ylidene]acetate
Formel: C₁₅H₁₅NO₃
Molecular Weight: 257,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568302
CAS-Nummer: 1308732-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR